The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193416.2 | NP_001180345.1 | 2 - 3 | 200 | 104 |
NM_001356.4 | NP_001347.3 | 2 - 3 | 1054 | 104 | |
NR_126093.1 | 2 - 3 | 1054 | 104 | ||
XM_011543892.2 | XP_011542194.1 | 2 - 3 | 231 | 104 | |
GenBank mRNA | AB208983.1 | - | 2 - 3 | 152 | 104 |
AB451220.1 | - | 2 - 3 | 109 | 104 | |
AB451343.1 | - | 2 - 3 | 109 | 104 | |
AF061337.1 | - | 2 - 3 | 184 | 104 | |
AK291153.1 | - | 2 - 3 | 200 | 104 | |
AK296906.1 | - | 2 - 3 | 200 | 104 | |
AK297159.1 | - | 2 - 3 | 200 | 104 | |
AK304661.1 | - | 2 - 3 | 243 | 104 | |
BC011819.2 | - | 2 - 3 | 179 | 104 | |
BM833870.1 | - | 2 - 3 | 200 | 104 | |
U50553.1 | - | 2 - 3 | 418 | 104 |