The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000097.5 | NP_000088.3 | 2 - 3 | 809 | 77 |
XM_005247125.4 | XP_005247182.1 | 2 - 3 | 873 | 77 | |
GenBank mRNA | AK223481.1 | - | 2 - 3 | 739 | 77 |
AK290140.1 | - | 2 - 3 | 809 | 77 | |
AK299692.1 | - | 2 - 3 | 811 | 77 | |
BC017210.2 | - | 2 - 3 | 760 | 77 | |
BC023551.2 | - | 2 - 3 | 795 | 77 | |
BC023554.2 | - | 2 - 3 | 756 | 77 | |
D16611.1 | - | 2 - 3 | 495 | 77 | |
Z28409.1 | - | 2 - 3 | 402 | 77 |