Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC1
Entrez Gene ID
55227
Gene Name
leucine rich repeat containing 1
Gene Aliases
LANO, dJ523E19.1
Chromosome Location
Chr.6: 53794497 - 53924125 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018214.4 NP_060684.4 9 - 10 1182 127
XM_011514726.2 XP_011513028.1 10 - 11 1546 127
XM_011514727.2 XP_011513029.1 8 - 9 972 127
XM_017010997.1 XP_016866486.1 9 - 10 1474 127
GenBank mRNA AF332199.1 - 9 - 10 1129 127
AF359380.1 - 9 - 10 905 127
AK001637.1 - 9 - 10 885 127
AK021896.1 - 10 - 11 1542 127
AK292248.1 - 9 - 10 1027 127
BC003193.1 - 9 - 10 1182 127

View Full Product Details