The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001431.3 | NP_001422.1 | 15 - 16 | 2789 | 90 |
XM_006715362.2 | XP_006715425.1 | 13 - 14 | 2610 | 90 | |
XM_011535524.1 | XP_011533826.1 | 17 - 18 | 2945 | 90 | |
XM_011535525.1 | XP_011533827.1 | 16 - 17 | 2891 | 90 | |
XM_011535527.1 | XP_011533829.1 | 16 - 17 | 2843 | 90 | |
XM_011535528.1 | XP_011533830.1 | 14 - 15 | 2633 | 90 | |
XM_011535529.1 | XP_011533831.1 | 13 - 14 | 2579 | 90 | |
XM_017010349.1 | XP_016865838.1 | 17 - 18 | 2973 | 90 | |
XM_017010350.1 | XP_016865839.1 | 17 - 18 | 2907 | 90 | |
XM_017010351.1 | XP_016865840.1 | 17 - 18 | 2969 | 90 | |
XM_017010352.1 | XP_016865841.1 | 17 - 18 | 2811 | 90 | |
XM_017010353.1 | XP_016865842.1 | 17 - 18 | 2945 | 90 | |
XM_017010354.1 | XP_016865843.1 | 16 - 17 | 2882 | 90 | |
XM_017010355.1 | XP_016865844.1 | 15 - 16 | 2817 | 90 | |
XM_017010357.1 | XP_016865846.1 | 13 - 14 | 2541 | 90 | |
XM_017010364.1 | XP_016865853.1 | 15 - 16 | 2194 | 90 | |
GenBank mRNA | AF027299.1 | - | 15 - 16 | 2651 | 90 |
AY047584.1 | - | 14 - 15 | 2615 | 90 | |
AY512660.1 | - | 4 - 5 | 2210 | 90 |