Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EPB41L2
- Entrez Gene ID
-
2037
- Gene Name
- erythrocyte membrane protein band 4.1 like 2
- Gene Aliases
-
4.1-G, 4.1G
- Chromosome Location
-
Chr.6: 130829111 - 131063322 on Build GRCh38
View Full Product Details