The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004133.4 | NP_004124.4 | 4 - 5 | 481 | 58 |
XM_011517516.2 | XP_011515818.1 | 6 - 7 | 918 | 58 | |
XM_017013373.1 | XP_016868862.1 | 7 - 8 | 996 | 58 | |
XM_017013374.1 | XP_016868863.1 | 5 - 6 | 886 | 58 | |
XM_017013375.1 | XP_016868864.1 | 6 - 7 | 663 | 58 | |
XM_017013376.1 | XP_016868865.1 | 5 - 6 | 511 | 58 | |
GenBank mRNA | AB307704.1 | - | 3 - 4 | 345 | 58 |
BC105009.1 | - | 3 - 4 | 375 | 58 | |
BC105011.1 | - | 3 - 4 | 375 | 58 | |
BX571750.1 | - | 4 - 5 | 481 | 58 | |
CR542091.1 | - | 3 - 4 | 345 | 58 | |
HQ692842.1 | - | 4 - 5 | 456 | 58 | |
Z49826.2 | - | 5 - 6 | 615 | 58 |