Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRPF1
Entrez Gene ID
7862
Gene Name
bromodomain and PHD finger containing 1
Gene Aliases
BR140
Chromosome Location
Chr.3: 9731729 - 9748015 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003694.1 NP_001003694.1 10 - 11 3467 69
NM_001319049.1 NP_001305978.1 9 - 10 3185 69
NM_001319050.1 NP_001305979.1 10 - 11 3467 69
NM_004634.2 NP_004625.2 10 - 11 3449 69
XM_005265449.1 XP_005265506.1 10 - 11 3467 69
XM_005265450.1 XP_005265507.1 10 - 11 3485 69
XM_005265452.1 XP_005265509.1 9 - 10 3203 69
XM_011534101.1 XP_011532403.1 10 - 11 3470 69
XM_011534102.1 XP_011532404.1 10 - 11 3268 69
GenBank mRNA AK293865.1 - 9 - 10 3185 69
AL713696.1 - 10 - 11 3276 69
BC053851.1 - 10 - 11 3461 69
M91585.1 - 10 - 11 3078 69

View Full Product Details