The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001003694.1 | NP_001003694.1 | 12 - 13 | 3721 | 81 |
NM_001319049.1 | NP_001305978.1 | 11 - 12 | 3439 | 81 | |
NM_001319050.1 | NP_001305979.1 | 12 - 13 | 3721 | 81 | |
NM_004634.2 | NP_004625.2 | 12 - 13 | 3703 | 81 | |
XM_005265449.1 | XP_005265506.1 | 11 - 12 | 3823 | 81 | |
XM_005265450.1 | XP_005265507.1 | 12 - 13 | 3739 | 81 | |
XM_005265452.1 | XP_005265509.1 | 11 - 12 | 3457 | 81 | |
XM_011534101.1 | XP_011532403.1 | 11 - 12 | 3826 | 81 | |
XM_011534102.1 | XP_011532404.1 | 11 - 12 | 3624 | 81 | |
GenBank mRNA | AK293865.1 | - | 11 - 12 | 3439 | 81 |
AL713696.1 | - | 12 - 13 | 3530 | 81 | |
BC053851.1 | - | 12 - 13 | 3715 | 81 | |
M91585.1 | - | 12 - 13 | 3332 | 81 |