The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039481.1 | NP_001034570.1 | 1 - 2 | 443 | 90 |
NM_018638.4 | NP_061108.2 | 1 - 2 | 443 | 90 | |
XM_005253420.4 | XP_005253477.1 | 1 - 2 | 440 | 90 | |
XM_017019580.1 | XP_016875069.1 | 1 - 2 | 440 | 90 | |
XM_017019581.1 | XP_016875070.1 | 1 - 2 | 440 | 90 | |
GenBank mRNA | AF207600.2 | - | 1 - 2 | 509 | 90 |
AK074425.1 | - | 1 - 2 | 388 | 90 | |
AL137750.1 | - | 1 - 2 | 393 | 90 | |
BC006111.1 | - | 1 - 2 | 395 | 90 | |
BC066907.1 | - | 1 - 2 | 443 | 90 | |
BT007084.1 | - | 1 - 2 | 154 | 90 |