Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ETNK1
Entrez Gene ID
55500
Gene Name
ethanolamine kinase 1
Gene Aliases
EKI, EKI 1, EKI1, Nbla10396
Chromosome Location
Chr.12: 22625142 - 22690674 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018638.4 NP_061108.2 3 - 4 847 110
XM_017019580.1 XP_016875069.1 3 - 4 844 110
XM_017019581.1 XP_016875070.1 3 - 4 844 110
GenBank mRNA AF207600.2 - 3 - 4 913 110
AK074425.1 - 3 - 4 792 110
AL137750.1 - 3 - 4 796 110
BC066907.1 - 3 - 4 847 110

View Full Product Details