The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018040.3 | NP_060510.1 | 9 - 10 | 1495 | 70 |
XM_017001592.1 | XP_016857081.1 | 7 - 8 | 1594 | 70 | |
GenBank mRNA | AK001114.1 | - | 9 - 10 | 1462 | 70 |
AK024701.1 | - | 2 - 3 | 135 | 70 |