The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015897.3 | NP_056981.2 | 8 - 9 | 1146 | 82 |
XM_011528060.2 | XP_011526362.1 | 8 - 9 | 1489 | 82 | |
XM_017026868.1 | XP_016882357.1 | 7 - 8 | 966 | 82 | |
GenBank mRNA | AF077952.1 | - | 8 - 9 | 983 | 82 |
AF164437.1 | - | 7 - 8 | 956 | 82 | |
AK022481.1 | - | 8 - 9 | 994 | 82 | |
AK222726.1 | - | 8 - 9 | 986 | 82 | |
AK315530.1 | - | 8 - 9 | 1026 | 82 | |
BC004389.1 | - | 4 - 5 | 396 | 82 | |
BC010047.2 | - | 8 - 9 | 973 | 82 | |
BC029874.1 | - | 8 - 9 | 993 | 82 | |
BC066895.1 | - | 8 - 9 | 996 | 82 |