Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KHK
Entrez Gene ID
3795
Gene Name
ketohexokinase
Gene Aliases
-
Chromosome Location
Chr.2: 27086738 - 27100751 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000221.2 NP_000212.1 7 - 8 1325 66
NM_006488.2 NP_006479.1 7 - 8 1325 66
XM_005264294.3 XP_005264351.1 8 - 9 1468 66
XM_005264296.3 XP_005264353.1 7 - 8 1311 66
XM_005264298.3 XP_005264355.1 5 - 6 1078 66
XM_017004060.1 XP_016859549.1 8 - 9 1465 66
XM_017004061.1 XP_016859550.1 6 - 7 1195 66
GenBank mRNA AK130033.1 - 7 - 8 842 66
AK223075.1 - 7 - 8 842 66
BC006233.2 - 7 - 8 1263 66
BX648873.1 - 5 - 6 592 66
CR456801.1 - 7 - 8 808 66
X78677.1 - 5 - 6 604 66
X78678.1 - 7 - 8 816 66

View Full Product Details