Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CADPS
Entrez Gene ID
8618
Gene Name
calcium dependent secretion activator
Gene Aliases
CADPS1, CAPS, CAPS1, UNC-31
Chromosome Location
Chr.3: 62398346 - 62875389 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003716.3 NP_003707.2 14 - 15 2751 74
NM_183393.2 NP_899630.1 14 - 15 2700 74
NM_183394.2 NP_899631.1 14 - 15 2751 74
XM_006713378.2 XP_006713441.2 15 - 16 2758 74
XM_011534177.1 XP_011532479.1 16 - 17 2917 74
XM_011534178.1 XP_011532480.1 16 - 17 2917 74
XM_011534180.1 XP_011532482.1 16 - 17 2917 74
XM_011534182.1 XP_011532484.1 16 - 17 2917 74
XM_011534183.1 XP_011532485.1 16 - 17 2917 74
XM_011534189.1 XP_011532491.1 16 - 17 2917 74
XM_011534191.2 XP_011532493.1 14 - 15 2391 74
XM_011534192.1 XP_011532494.1 16 - 17 2917 74
XM_011534193.1 XP_011532495.1 15 - 16 2758 74
XM_011534194.2 XP_011532496.1 15 - 16 2403 74
XM_011534195.1 XP_011532497.1 16 - 17 2917 74
XM_011534196.1 XP_011532498.1 15 - 16 2758 74
XM_011534197.2 XP_011532499.1 15 - 16 2863 74
XM_011534198.2 XP_011532500.1 14 - 15 2391 74
XM_011534199.2 XP_011532501.1 14 - 15 2391 74
XM_011534200.2 XP_011532502.1 14 - 15 2391 74
XM_011534202.2 XP_011532504.1 16 - 17 2917 74
XM_011534203.1 XP_011532505.1 15 - 16 2758 74
XM_017007358.1 XP_016862847.1 16 - 17 2917 74
XM_017007359.1 XP_016862848.1 16 - 17 2917 74
XM_017007360.1 XP_016862849.1 16 - 17 2917 74
XM_017007361.1 XP_016862850.1 16 - 17 2917 74
XM_017007362.1 XP_016862851.1 16 - 17 2917 74
XM_017007363.1 XP_016862852.1 16 - 17 2917 74
XM_017007364.1 XP_016862853.1 16 - 17 2917 74
XM_017007365.1 XP_016862854.1 15 - 16 2758 74
XM_017007366.1 XP_016862855.1 16 - 17 2917 74
XM_017007367.1 XP_016862856.1 14 - 15 2391 74
XM_017007368.1 XP_016862857.1 14 - 15 2391 74
XM_017007369.1 XP_016862858.1 15 - 16 2758 74
XM_017007370.1 XP_016862859.1 16 - 17 2917 74
XM_017007371.1 XP_016862860.1 16 - 17 2917 74
XM_017007372.1 XP_016862861.1 14 - 15 2391 74
XM_017007373.1 XP_016862862.1 15 - 16 2758 74
XM_017007374.1 XP_016862863.1 15 - 16 2758 74
XM_017007375.1 XP_016862864.1 14 - 15 2391 74
XM_017007376.1 XP_016862865.1 16 - 17 2917 74
XM_017007377.1 XP_016862866.1 14 - 15 2391 74
XM_017007378.1 XP_016862867.1 14 - 15 2391 74
XM_017007379.1 XP_016862868.1 15 - 16 2758 74
XM_017007380.1 XP_016862869.1 15 - 16 2758 74
XM_017007381.1 XP_016862870.1 15 - 16 2758 74
XM_017007382.1 XP_016862871.1 14 - 15 2391 74
XM_017007383.1 XP_016862872.1 15 - 16 2758 74
XM_017007384.1 XP_016862873.1 15 - 16 2758 74
XM_017007385.1 XP_016862874.1 15 - 16 2758 74
XM_017007386.1 XP_016862875.1 15 - 16 2758 74
GenBank mRNA AB032947.2 - 14 - 15 2741 74
AB593079.1 - 15 - 16 2863 74
AF458662.1 - 14 - 15 2650 74
AL833895.1 - 14 - 15 2048 74
BC131731.1 - 7 - 8 877 74

View Full Product Details