The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005506.3 | NP_005497.1 | 5 - 6 | 1064 | 160 |
GenBank mRNA | AK313016.1 | - | 5 - 6 | 774 | 160 |
BC021892.1 | - | 5 - 6 | 730 | 160 | |
BT006939.1 | - | 5 - 6 | 708 | 160 | |
D12676.1 | - | 5 - 6 | 959 | 160 |