Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCARB2
Entrez Gene ID
950
Gene Name
scavenger receptor class B member 2
Gene Aliases
AMRF, CD36L2, EPM4, HLGP85, LGP85, LIMP-2, LIMPII, SR-BII
Chromosome Location
Chr.4: 76158737 - 76213899 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204255.1 NP_001191184.1 3 - 4 747 78
NM_005506.3 NP_005497.1 6 - 7 1176 78
GenBank mRNA AK296519.1 - 3 - 4 451 78
AK313016.1 - 6 - 7 886 78
BC021892.1 - 6 - 7 842 78
BT006939.1 - 6 - 7 820 78
D12676.1 - 6 - 7 1071 78

View Full Product Details