The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258392.2 | NP_001245321.1 | 11 - 12 | 1540 | 65 |
NM_001258393.2 | NP_001245322.1 | 10 - 11 | 1453 | 65 | |
NM_001258394.2 | NP_001245323.1 | 13 - 14 | 1737 | 65 | |
NM_030813.5 | NP_110440.1 | 12 - 13 | 1630 | 65 | |
XM_005274320.1 | XP_005274377.1 | 11 - 12 | 1416 | 65 | |
GenBank mRNA | AK023214.1 | - | 11 - 12 | 1390 | 65 |
AK302006.1 | - | 10 - 11 | 1296 | 65 | |
AK302069.1 | - | 13 - 14 | 1435 | 65 | |
AK302158.1 | - | 10 - 11 | 1371 | 65 | |
AL136909.1 | - | 12 - 13 | 1529 | 65 | |
AL834484.1 | - | 3 - 4 | 265 | 65 | |
BC006257.2 | - | 4 - 5 | 347 | 65 | |
BC006404.2 | - | 12 - 13 | 1470 | 65 |