The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258392.2 | NP_001245321.1 | 15 - 16 | 2001 | 55 |
NM_001258393.2 | NP_001245322.1 | 14 - 15 | 1914 | 55 | |
NM_001258394.2 | NP_001245323.1 | 17 - 18 | 2198 | 55 | |
NM_030813.5 | NP_110440.1 | 16 - 17 | 2091 | 55 | |
XM_005274320.1 | XP_005274377.1 | 15 - 16 | 1877 | 55 | |
GenBank mRNA | AK023214.1 | - | 15 - 16 | 1851 | 55 |
AK302006.1 | - | 14 - 15 | 1757 | 55 | |
AK302069.1 | - | 17 - 18 | 1896 | 55 | |
AK302158.1 | - | 14 - 15 | 1832 | 55 | |
AL136909.1 | - | 16 - 17 | 1990 | 55 | |
AL834484.1 | - | 7 - 8 | 726 | 55 | |
BC006257.2 | - | 8 - 9 | 808 | 55 | |
BC006404.2 | - | 16 - 17 | 1931 | 55 |