The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021957.3 | NP_068776.2 | 9 - 10 | 1489 | 156 |
XM_006719063.3 | XP_006719126.1 | 8 - 9 | 1021 | 156 | |
GenBank mRNA | AK308373.1 | - | 9 - 10 | 1461 | 156 |
AV653197.1 | - | 2 - 3 | 156 | ||
BC126310.1 | - | 9 - 10 | 1259 | 156 | |
BC126312.1 | - | 9 - 10 | 1259 | 156 | |
D29685.1 | - | 9 - 10 | 1491 | 156 | |
S70004.1 | - | 9 - 10 | 1262 | 156 |