Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHIT1
Entrez Gene ID
1118
Gene Name
chitinase 1
Gene Aliases
CHI3, CHIT, CHITD
Chromosome Location
Chr.1: 203216079 - 203229732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256125.1 NP_001243054.2 7 - 8 960 78
NM_001270509.1 NP_001257438.1 8 - 9 969 78
NM_003465.2 NP_003456.1 8 - 9 1017 78
NR_045784.1 8 - 9 1017 78
NR_045785.1 8 - 9 1017 78
GenBank mRNA AK055165.1 - 7 - 8 900 78
AK131093.1 - 9 - 10 5152 78
AK225245.1 - 8 - 9 909 78
AK309341.1 - 8 - 9 959 78
AK314331.1 - 8 - 9 956 78
BC069614.1 - 7 - 8 886 78
BC103695.1 - 8 - 9 933 78
BC105680.1 - 8 - 9 933 78
BC105681.1 - 8 - 9 933 78
BC105682.1 - 8 - 9 933 78
BP297140.1 - 3 - 4 355 78
U29615.1 - 8 - 9 933 78
U62662.1 - 8 - 9 930 78

View Full Product Details