Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATN1
Entrez Gene ID
1822
Gene Name
atrophin 1
Gene Aliases
B37, D12S755E, DRPLA, HRS, NOD
Chromosome Location
Chr.12: 6924463 - 6942321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BE142829.1 - 4 - 5 536 71
U23851.1 - 3 - 4 71

View Full Product Details