The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 11 - 12 | 1577 | 106 |
XM_006715130.2 | XP_006715193.1 | 11 - 12 | 1543 | 106 | |
XM_011514722.1 | XP_011513024.1 | 12 - 13 | 1567 | 106 | |
XM_017010995.1 | XP_016866484.1 | 10 - 11 | 1712 | 106 | |
GenBank mRNA | AJ544537.1 | - | 11 - 12 | 1510 | 106 |
BC136294.1 | - | 10 - 11 | 1161 | 106 | |
Y13247.1 | - | 10 - 11 | 1500 | 106 |