The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 18 - 19 | 2575 | 99 |
NR_072994.1 | 18 - 19 | 2514 | 99 | ||
XM_006715130.2 | XP_006715193.1 | 18 - 19 | 2541 | 99 | |
XM_011514722.1 | XP_011513024.1 | 19 - 20 | 2565 | 99 | |
XM_017010995.1 | XP_016866484.1 | 17 - 18 | 2710 | 99 | |
GenBank mRNA | AJ544537.1 | - | 18 - 19 | 2508 | 99 |
BC136294.1 | - | 17 - 18 | 2159 | 99 | |
BC150310.1 | - | 18 - 19 | 2515 | 99 | |
Y13247.1 | - | 17 - 18 | 2498 | 99 |