The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 5 - 6 | 945 | 66 |
NR_072994.1 | 5 - 6 | 945 | 66 | ||
XM_006715130.2 | XP_006715193.1 | 5 - 6 | 911 | 66 | |
XM_011514722.1 | XP_011513024.1 | 6 - 7 | 935 | 66 | |
XM_017010995.1 | XP_016866484.1 | 4 - 5 | 1080 | 66 | |
GenBank mRNA | AJ544537.1 | - | 5 - 6 | 878 | 66 |
AK307406.1 | - | 5 - 6 | 881 | 66 | |
BC005267.2 | - | 4 - 5 | 862 | 66 | |
BC136294.1 | - | 4 - 5 | 529 | 66 | |
BC150310.1 | - | 5 - 6 | 946 | 66 |