The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002714.3 | NP_002705.2 | 7 - 8 | 1073 | 93 |
NR_072994.1 | 7 - 8 | 1073 | 93 | ||
XM_006715130.2 | XP_006715193.1 | 7 - 8 | 1039 | 93 | |
XM_011514722.1 | XP_011513024.1 | 8 - 9 | 1063 | 93 | |
XM_017010995.1 | XP_016866484.1 | 6 - 7 | 1208 | 93 | |
GenBank mRNA | AJ544537.1 | - | 7 - 8 | 1006 | 93 |
AK307406.1 | - | 7 - 8 | 1009 | 93 | |
BC005267.2 | - | 6 - 7 | 990 | 93 | |
BC136294.1 | - | 6 - 7 | 657 | 93 | |
BC150310.1 | - | 7 - 8 | 1074 | 93 | |
Y13247.1 | - | 6 - 7 | 996 | 93 |