Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP85
Entrez Gene ID
64793
Gene Name
centrosomal protein 85
Gene Aliases
CCDC21
Chromosome Location
Chr.1: 26234153 - 26278810 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281517.2 NP_001268446.1 7 - 8 1521 68
NM_001319944.1 NP_001306873.1 8 - 9 1674 68
NM_022778.4 NP_073615.2 8 - 9 1674 68
XM_017002105.1 XP_016857594.1 8 - 9 1674 68
GenBank mRNA AK024038.1 - 8 - 9 1630 68
AK025653.1 - 2 - 3 273 68
AK299312.1 - 7 - 8 1474 68
AL133609.1 - 4 - 5 515 68
AL137420.1 - 2 - 3 272 68
BC064528.1 - 8 - 9 1625 68

View Full Product Details