Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX3X
Entrez Gene ID
1654
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CAP-Rf, DBX, DDX14, DDX3, HLP2, MRX102
Chromosome Location
Chr.X: 41333308 - 41364472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_126093.1 17 - 18 3127 84
GenBank mRNA BF980241.1 - 2 - 3 84

View Full Product Details