The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005502.2 | NP_001005502.1 | 8 - 9 | 1138 | 134 |
NM_001874.4 | NP_001865.1 | 8 - 9 | 1170 | 134 | |
NM_198320.3 | NP_938079.1 | 8 - 9 | 1156 | 134 | |
GenBank mRNA | AF368463.1 | - | 8 - 9 | 134 | |
AK130146.1 | - | 5 - 6 | 746 | 134 | |
AK313180.1 | - | 8 - 9 | 1128 | 134 | |
BC022276.1 | - | 8 - 9 | 1128 | 134 | |
J04970.2 | - | 8 - 9 | 134 |