The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001046.2 | NP_001037.1 | 12 - 13 | 2169 | 146 |
NM_001256461.1 | NP_001243390.1 | 12 - 13 | 2169 | 146 | |
NR_046207.1 | 12 - 13 | 2169 | 146 | ||
XM_017009771.1 | XP_016865260.1 | 11 - 12 | 1319 | 146 | |
GenBank mRNA | AY280459.1 | - | 12 - 13 | 2194 | 146 |
BC144221.1 | - | 12 - 13 | 2112 | 146 | |
BC146839.1 | - | 12 - 13 | 2112 | 146 | |
U30246.1 | - | 12 - 13 | 2169 | 146 |