The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001046.2 | NP_001037.1 | 8 - 9 | 1702 | 133 |
NM_001256461.1 | NP_001243390.1 | 8 - 9 | 1702 | 133 | |
NR_046207.1 | 8 - 9 | 1702 | 133 | ||
XM_011543588.2 | XP_011541890.1 | 8 - 9 | 1696 | 133 | |
GenBank mRNA | AY280459.1 | - | 8 - 9 | 1727 | 133 |
BC144221.1 | - | 8 - 9 | 1645 | 133 | |
BC146839.1 | - | 8 - 9 | 1645 | 133 | |
U30246.1 | - | 8 - 9 | 1702 | 133 |