Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMA1
Entrez Gene ID
284217
Gene Name
laminin subunit alpha 1
Gene Aliases
LAMA, PTBHS, S-LAM-alpha
Chromosome Location
Chr.18: 6941744 - 7117814 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005559.3 NP_005550.2 50 - 51 7295 87
XM_011525655.2 XP_011523957.1 50 - 51 7289 87
XM_011525656.2 XP_011523958.1 40 - 41 5710 87
XM_011525657.1 XP_011523959.1 50 - 51 7273 87
GenBank mRNA AK091949.1 - 1 - 2 675 87
BC039051.1 - 2 - 3 219 87
BC144590.1 - 49 - 50 8216 87
X58531.1 - 50 - 51 7295 87

View Full Product Details