Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CCDC169-SOHLH2
- Entrez Gene ID
-
100526761
- Gene Name
- CCDC169-SOHLH2 readthrough
- Gene Aliases
-
C13orf38-SOHLH2
- Chromosome Location
-
Chr.13: 36168208 - 36297855 on Build GRCh38
- Gene Symbol
- SOHLH2
- Entrez Gene ID
-
54937
- Gene Name
- spermatogenesis and oogenesis specific basic helix-loop-helix 2
- Gene Aliases
-
SPATA28, TEB1, bHLHe81
- Chromosome Location
-
Chr.13: 36168208 - 36214615 on Build GRCh38
View Full Product Details