Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CUBN
Entrez Gene ID
8029
Gene Name
cubilin
Gene Aliases
IFCR, MGA1, gp280
Chromosome Location
Chr.10: 16823966 - 17130492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081.3 NP_001072.2 57 - 58 9161 99
XM_011519709.2 XP_011518011.1 31 - 32 5524 99
XM_011519710.2 XP_011518012.1 31 - 32 5167 99
XM_011519711.2 XP_011518013.1 30 - 31 5848 99
GenBank mRNA AB209880.1 - 30 - 31 5227 99
AF034611.1 - 57 - 58 9135 99
AK308290.1 - 9 - 10 1473 99

View Full Product Details