The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282755.1 | NP_001269684.1 | 8 - 9 | 1102 | 64 |
NM_001282756.1 | NP_001269685.1 | 7 - 8 | 754 | 64 | |
NM_020744.3 | NP_065795.1 | 7 - 8 | 829 | 64 | |
XM_005264456.2 | XP_005264513.1 | 7 - 8 | 631 | 64 | |
XM_005264458.2 | XP_005264515.1 | 7 - 8 | 631 | 64 | |
XM_005264459.2 | XP_005264516.1 | 8 - 9 | 1102 | 64 | |
XM_017004561.1 | XP_016860050.1 | 7 - 8 | 631 | 64 | |
XM_017004562.1 | XP_016860051.1 | 7 - 8 | 631 | 64 | |
GenBank mRNA | AB033092.1 | - | 7 - 8 | 754 | 64 |
AK027304.1 | - | 7 - 8 | 627 | 64 | |
AK127245.1 | - | 8 - 9 | 1102 | 64 | |
BC004227.2 | - | 7 - 8 | 657 | 64 | |
BC053631.1 | - | 7 - 8 | 815 | 64 |