Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL35
Entrez Gene ID
283212
Gene Name
kelch like family member 35
Gene Aliases
-
Chromosome Location
Chr.11: 75422394 - 75434550 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039548.2 NP_001034637.2 4 - 5 1373 127
XM_011544954.2 XP_011543256.1 5 - 6 2882 127
GenBank mRNA AK091109.1 - 3 - 4 1768 127
BC042952.2 - 4 - 5 914 127
BC057763.1 - 4 - 5 935 127
BC131539.1 - 1 - 2 354 127
BC132708.1 - 4 - 5 737 127
BC132710.1 - 4 - 5 737 127

View Full Product Details