The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014429.3 | NP_055244.3 | 27 - 28 | 2883 | 66 |
XM_005247362.2 | XP_005247419.1 | 28 - 29 | 2930 | 66 | |
XM_011512691.1 | XP_011510993.1 | 28 - 29 | 2936 | 66 | |
XM_011512692.1 | XP_011510994.1 | 27 - 28 | 2846 | 66 | |
XM_011512694.1 | XP_011510996.1 | 25 - 26 | 2687 | 66 | |
XM_011512696.2 | XP_011510998.1 | 22 - 23 | 2387 | 66 | |
XM_011512697.2 | XP_011510999.1 | 21 - 22 | 2555 | 66 | |
XM_017006170.1 | XP_016861659.1 | 20 - 21 | 2265 | 66 | |
GenBank mRNA | AF084946.1 | - | 27 - 28 | 2858 | 66 |
AK302642.1 | - | 26 - 27 | 2777 | 66 | |
AL353955.1 | - | 21 - 22 | 2243 | 66 | |
BC028680.1 | - | 27 - 28 | 2884 | 66 | |
BC050307.1 | - | 27 - 28 | 2822 | 66 |