Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIAA1549L
Entrez Gene ID
25758
Gene Name
KIAA1549 like
Gene Aliases
C11orf41, C11orf69, G2
Chromosome Location
Chr.11: 33376569 - 33674102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012194.2 NP_036326.2 10 - 11 3801 63
XM_005252847.3 XP_005252904.2 12 - 13 3895 63
XM_005252848.3 XP_005252905.1 10 - 11 3819 63
XM_011519970.2 XP_011518272.1 11 - 12 2461 63
XM_017017484.1 XP_016872973.1 10 - 11 3819 63
XM_017017485.1 XP_016872974.1 10 - 11 3819 63
XM_017017486.1 XP_016872975.1 11 - 12 2461 63
GenBank mRNA U10991.1 - 10 - 11 3208 63

View Full Product Details