The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000289.5 | NP_000280.1 | 15 - 16 | 1590 | 76 |
NM_001166686.1 | NP_001160158.1 | 17 - 18 | 1892 | 76 | |
NM_001166687.1 | NP_001160159.1 | 15 - 16 | 1444 | 76 | |
NM_001166688.1 | NP_001160160.1 | 14 - 15 | 1533 | 76 | |
XM_005268974.1 | XP_005269031.1 | 18 - 19 | 1789 | 76 | |
XM_005268975.1 | XP_005269032.1 | 18 - 19 | 1928 | 76 | |
XM_005268976.3 | XP_005269033.1 | 18 - 19 | 1971 | 76 | |
XM_005268977.1 | XP_005269034.1 | 17 - 18 | 1775 | 76 | |
XM_005268978.2 | XP_005269035.1 | 17 - 18 | 1688 | 76 | |
XM_005268979.1 | XP_005269036.1 | 17 - 18 | 1775 | 76 | |
XM_011538487.1 | XP_011536789.1 | 16 - 17 | 1630 | 76 | |
XM_011538488.2 | XP_011536790.1 | 15 - 16 | 1555 | 76 | |
XM_017019467.1 | XP_016874956.1 | 18 - 19 | 1858 | 76 | |
XM_017019468.1 | XP_016874957.1 | 17 - 18 | 1710 | 76 | |
XM_017019469.1 | XP_016874958.1 | 16 - 17 | 1682 | 76 | |
GenBank mRNA | AK126229.1 | - | 18 - 19 | 1842 | 76 |
AK303681.1 | - | 10 - 11 | 937 | 76 | |
AK308185.1 | - | 10 - 11 | 913 | 76 | |
AK312284.1 | - | 15 - 16 | 1472 | 76 | |
BC000534.2 | - | 14 - 15 | 1420 | 76 | |
BC007798.2 | - | 13 - 14 | 1307 | 76 | |
BC012799.2 | - | 14 - 15 | 1420 | 76 | |
BC013298.2 | - | 14 - 15 | 1420 | 76 | |
BC021203.2 | - | 15 - 16 | 1442 | 76 | |
CR541698.1 | - | 5 - 6 | 460 | 76 | |
J05533.1 | - | 6 - 7 | 501 | 76 | |
KU178251.1 | - | 14 - 15 | 1405 | 76 | |
KU178252.1 | - | 13 - 14 | 1312 | 76 | |
KU178253.1 | - | 14 - 15 | 1405 | 76 | |
KU178254.1 | - | 3 - 4 | 250 | 76 | |
M26066.1 | - | 15 - 16 | 1481 | 76 | |
U24183.1 | - | 14 - 15 | 1733 | 76 | |
Y00698.1 | - | 14 - 15 | 1424 | 76 |