Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UVRAG
Entrez Gene ID
7405
Gene Name
UV radiation resistance associated
Gene Aliases
DHTX, VPS38, p63
Chromosome Location
Chr.11: 75814852 - 76144238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003369.3 NP_003360.2 5 - 6 751 87
XM_011545226.2 XP_011543528.1 5 - 6 1066 87
XM_011545227.2 XP_011543529.1 5 - 6 1066 87
XM_011545228.2 XP_011543530.1 5 - 6 1066 87
XM_017018226.1 XP_016873715.1 5 - 6 1066 87
GenBank mRNA AB012958.1 - 5 - 6 688 87
AK222772.1 - 5 - 6 642 87
AK291011.1 - 5 - 6 729 87
AK293782.1 - 5 - 6 492 87
BC064837.1 - 5 - 6 661 87

View Full Product Details