The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127384.2 | NP_001120856.1 | 14 - 15 | 2126 | 134 |
NM_013266.3 | NP_037398.2 | 14 - 15 | 2173 | 134 | |
XM_017016151.1 | XP_016871640.1 | 15 - 16 | 2168 | 134 | |
XM_017016152.1 | XP_016871641.1 | 17 - 18 | 2465 | 134 | |
XM_017016153.1 | XP_016871642.1 | 15 - 16 | 2176 | 134 | |
XM_017016154.1 | XP_016871643.1 | 10 - 11 | 1544 | 134 | |
XM_017016155.1 | XP_016871644.1 | 10 - 11 | 1554 | 134 | |
XM_017016156.1 | XP_016871645.1 | 10 - 11 | 1431 | 134 | |
XM_017016157.1 | XP_016871646.1 | 8 - 9 | 1581 | 134 | |
GenBank mRNA | AF091606.1 | - | 14 - 15 | 2151 | 134 |
AK289756.1 | - | 14 - 15 | 2062 | 134 | |
AK313882.1 | - | 14 - 15 | 2176 | 134 |