The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000026.3 | NP_000017.1 | 5 - 6 | 719 | 64 |
NM_001123378.2 | NP_001116850.1 | 5 - 6 | 719 | 64 | |
NM_001317923.1 | NP_001304852.1 | 5 - 6 | 664 | 64 | |
NR_134256.1 | 5 - 6 | 719 | 64 | ||
XM_011529977.2 | XP_011528279.1 | 5 - 6 | 719 | 64 | |
XM_011529979.1 | XP_011528281.1 | 5 - 6 | 719 | 64 | |
XM_011529980.2 | XP_011528282.1 | 5 - 6 | 719 | 64 | |
XM_017028636.1 | XP_016884125.1 | 4 - 5 | 656 | 64 | |
XM_017028637.1 | XP_016884126.1 | 4 - 5 | 639 | 64 | |
XM_017028638.1 | XP_016884127.1 | 3 - 4 | 576 | 64 | |
XM_017028639.1 | XP_016884128.1 | 3 - 4 | 576 | 64 | |
XM_017028640.1 | XP_016884129.1 | 1 - 2 | 74 | 64 | |
GenBank mRNA | AF067853.1 | - | 5 - 6 | 715 | 64 |
AF067854.1 | - | 5 - 6 | 715 | 64 | |
AK225042.1 | - | 5 - 6 | 687 | 64 | |
AK291091.1 | - | 5 - 6 | 690 | 64 | |
AK298187.1 | - | 5 - 6 | 733 | 64 | |
AK300705.1 | - | 3 - 4 | 496 | 64 | |
AK308662.1 | - | 1 - 2 | 74 | 64 | |
BC000253.1 | - | 5 - 6 | 689 | 64 | |
BC018881.2 | - | 5 - 6 | 689 | 64 | |
CR456368.1 | - | 5 - 6 | 693 | 64 | |
KJ534763.1 | - | 5 - 6 | 660 | 64 | |
KJ535007.1 | - | 5 - 6 | 660 | 64 | |
KJ535020.1 | - | 5 - 6 | 660 | 64 | |
KJ535041.1 | - | 2 - 3 | 331 | 64 | |
KJ535056.1 | - | 5 - 6 | 605 | 64 | |
S60710.1 | - | 5 - 6 | 662 | 64 | |
X65867.1 | - | 5 - 6 | 662 | 64 |