The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000895.2 | NP_000886.1 | 17 - 18 | 1756 | 83 |
NM_001256643.1 | NP_001243572.1 | 17 - 18 | 1687 | 83 | |
NR_132659.1 | 17 - 18 | 1756 | 83 | ||
GenBank mRNA | AB209740.1 | - | 17 - 18 | 5731 | 83 |
AK293630.1 | - | 17 - 18 | 1736 | 83 | |
AK301308.1 | - | 17 - 18 | 1565 | 83 | |
BC032528.1 | - | 17 - 18 | 1673 | 83 | |
BC036555.1 | - | 4 - 5 | 1505 | 83 | |
BX647158.1 | - | 17 - 18 | 1644 | 83 | |
CR457068.1 | - | 17 - 18 | 1614 | 83 | |
J02959.1 | - | 17 - 18 | 1644 | 83 | |
J03459.1 | - | 17 - 18 | 1682 | 83 |