The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267560.1 | NP_001254489.1 | 10 - 11 | 1362 | 83 |
NM_001267561.1 | NP_001254490.1 | 10 - 11 | 1349 | 83 | |
GenBank mRNA | AK027292.1 | - | 1 - 2 | 71 | 83 |
AK091118.1 | - | 10 - 11 | 1315 | 83 | |
AK128237.1 | - | 10 - 11 | 1311 | 83 | |
AK304462.1 | - | 9 - 10 | 1139 | 83 | |
BC108906.2 | - | 9 - 10 | 1185 | 83 | |
BC108907.1 | - | 9 - 10 | 1185 | 83 |