The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267560.1 | NP_001254489.1 | 4 - 5 | 502 | 67 |
NM_001267561.1 | NP_001254490.1 | 4 - 5 | 489 | 67 | |
GenBank mRNA | AK128237.1 | - | 4 - 5 | 451 | 67 |
AK304462.1 | - | 3 - 4 | 279 | 67 | |
BC108906.2 | - | 3 - 4 | 325 | 67 | |
BC108907.1 | - | 3 - 4 | 325 | 67 | |
DB079164.1 | - | 4 - 5 | 302 | 67 |