The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267560.1 | NP_001254489.1 | 6 - 7 | 953 | 73 |
NM_001267561.1 | NP_001254490.1 | 6 - 7 | 940 | 73 | |
GenBank mRNA | AK091118.1 | - | 6 - 7 | 906 | 73 |
AK128237.1 | - | 6 - 7 | 902 | 73 | |
AK304462.1 | - | 5 - 6 | 730 | 73 | |
BC108906.2 | - | 5 - 6 | 776 | 73 | |
BC108907.1 | - | 5 - 6 | 776 | 73 |