The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005228.3 | NP_005219.2 | 25 - 26 | 3360 | 92 |
GenBank mRNA | AB209442.1 | - | 25 - 26 | 3259 | 92 |
AK225422.1 | - | 23 - 24 | 3011 | 92 | |
AK290352.1 | - | 25 - 26 | 3160 | 92 | |
AK294750.1 | - | 25 - 26 | 3138 | 92 | |
AY550966.1 | - | 25 - 26 | 3160 | 92 | |
BC094761.1 | - | 24 - 25 | 3236 | 92 | |
DQ088980.1 | - | 25 - 26 | 3160 | 92 | |
GU255993.1 | - | 25 - 26 | 3157 | 92 | |
HQ912715.1 | - | 24 - 25 | 3225 | 92 | |
X00588.1 | - | 25 - 26 | 3300 | 92 |