The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005228.3 | NP_005219.2 | 5 - 6 | 875 | 57 |
NM_201282.1 | NP_958439.1 | 5 - 6 | 875 | 57 | |
NM_201283.1 | NP_958440.1 | 5 - 6 | 875 | 57 | |
NM_201284.1 | NP_958441.1 | 5 - 6 | 875 | 57 | |
GenBank mRNA | AB209442.1 | - | 5 - 6 | 774 | 57 |
AF125253.1 | - | 5 - 6 | 874 | 57 | |
AK225422.1 | - | 5 - 6 | 790 | 57 | |
AK290352.1 | - | 5 - 6 | 675 | 57 | |
AK294750.1 | - | 5 - 6 | 653 | 57 | |
AY550966.1 | - | 5 - 6 | 675 | 57 | |
AY698023.1 | - | 5 - 6 | 629 | 57 | |
AY698024.1 | - | 5 - 6 | 629 | 57 | |
BC094761.1 | - | 4 - 5 | 751 | 57 | |
DQ088980.1 | - | 5 - 6 | 675 | 57 | |
GU255993.1 | - | 5 - 6 | 672 | 57 | |
HQ912715.1 | - | 4 - 5 | 740 | 57 | |
K03193.1 | - | 5 - 6 | 788 | 57 | |
U48722.1 | - | 5 - 6 | 873 | 57 | |
U95089.1 | - | 6 - 7 | 1132 | 57 | |
X00588.1 | - | 5 - 6 | 815 | 57 | |
X00663.1 | - | 2 - 3 | 203 | 57 |