The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172648.3 | NP_001166119.1 | 5 - 6 | 1039 | 82 |
NM_001172649.3 | NP_001166120.1 | 5 - 6 | 1286 | 82 | |
NM_001291106.1 | NP_001278035.1 | 5 - 6 | 1286 | 82 | |
NM_001291107.1 | NP_001278036.1 | 5 - 6 | 1283 | 82 | |
NM_007185.6 | NP_009116.3 | 5 - 6 | 1286 | 82 | |
XM_006711139.2 | XP_006711202.1 | 6 - 7 | 514 | 82 | |
XM_006711141.2 | XP_006711204.1 | 6 - 7 | 511 | 82 | |
XM_011509111.2 | XP_011507413.1 | 5 - 6 | 1134 | 82 | |
XM_011509112.2 | XP_011507414.1 | 4 - 5 | 377 | 82 | |
XM_011509114.2 | XP_011507416.1 | 4 - 5 | 345 | 82 | |
XM_017000145.1 | XP_016855634.1 | 4 - 5 | 352 | 82 | |
XM_017000146.1 | XP_016855635.1 | 4 - 5 | 349 | 82 | |
GenBank mRNA | AB209264.1 | - | 5 - 6 | 1039 | 82 |
AF329264.1 | - | 5 - 6 | 486 | 82 | |
AL519587.3 | - | 6 - 7 | 518 | 82 | |
AY165003.1 | - | 5 - 6 | 1276 | 82 | |
BC004145.2 | - | 6 - 7 | 461 | 82 | |
BC041088.1 | - | 5 - 6 | 518 | 82 | |
BC052491.1 | - | 5 - 6 | 1260 | 82 | |
BC068008.1 | - | 4 - 5 | 313 | 82 | |
BC104758.1 | - | 5 - 6 | 549 | 82 | |
BC143226.1 | - | 5 - 6 | 546 | 82 | |
U80746.1 | - | 2 - 3 | 165 | 82 | |
U80759.1 | - | 2 - 3 | 172 | 82 |