The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005762.2 | NP_005753.1 | 14 - 15 | 2400 | 65 |
GenBank mRNA | AK131063.1 | - | 7 - 8 | 988 | 65 |
AK225760.1 | - | 14 - 15 | 2400 | 65 | |
AK313478.1 | - | 14 - 15 | 2117 | 65 | |
AL360114.1 | - | 2 - 3 | 226 | 65 | |
AL360188.1 | - | 2 - 3 | 307 | 65 | |
AL360189.1 | - | 2 - 3 | 187 | 65 | |
BC004978.1 | - | 14 - 15 | 2400 | 65 | |
BC007390.2 | - | 3 - 4 | 717 | 65 | |
BC052986.2 | - | 12 - 13 | 2151 | 65 | |
U31657.1 | - | 3 - 4 | 656 | 65 | |
U78773.1 | - | 14 - 15 | 2211 | 65 | |
U95040.1 | - | 14 - 15 | 2367 | 65 | |
X97548.1 | - | 14 - 15 | 2472 | 65 |