The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199135.1 | NP_001186064.1 | 4 - 5 | 432 | 119 |
NM_004180.2 | NP_004171.2 | 4 - 5 | 482 | 119 | |
XM_005246207.3 | XP_005246264.2 | 4 - 5 | 470 | 119 | |
XM_017003093.1 | XP_016858582.1 | 5 - 6 | 663 | 119 | |
XM_017003094.1 | XP_016858583.1 | 6 - 7 | 1005 | 119 | |
XM_017003095.1 | XP_016858584.1 | 6 - 7 | 910 | 119 | |
XM_017003096.1 | XP_016858585.1 | 4 - 5 | 464 | 119 | |
XM_017003097.1 | XP_016858586.1 | 6 - 7 | 738 | 119 | |
GenBank mRNA | AK313096.1 | - | 4 - 5 | 432 | 119 |
BC067779.1 | - | 4 - 5 | 424 | 119 | |
U59863.1 | - | 4 - 5 | 482 | 119 | |
U63830.1 | - | 4 - 5 | 481 | 119 |